Learn More
Cell Biolabs Inc PureVirus™ Adenovirus Purification Kit, 10 preps
Catalog No. NC1940168
Encompass_Preferred
Cell Biolabs' PureVirus™ Adenovirus Purification Kit is an efficient system for the purification of recombinant adenovirus preps, without the need for ultracentrifugation. The kit's purification resin quickly removes >98% of host cell proteins, nucleic acids, and other contaminants, with >60% virus recovery. The complete protocol takes less than two hours. Each prep can purify adenovirus harvested from up to four T75 flasks or four 10cm culture dishes.
Catalog No. NC1940168 Supplier Cell Biolabs Inc Supplier No. VPK5112
Cell Biolabs' PureVirus™ Adenovirus Purification Kit is an efficient system for the purification of recombinant adenovirus preps, without the need for ultracentrifugation. The kit's purification resin quickly removes >98% of host cell proteins, nucleic acids, and other contaminants, with >60% virus recovery. The complete protocol takes less than two hours. Each prep can purify adenovirus harvested from up to four T75 flasks or four 10cm culture dishes.