Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ Purewipes
SDP

Catalog No. 18444049
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18-444-049 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-049 Supplier Gentex™ Supplier No. PR02054SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Purewipes moist towelettes that clean dirt and perspiration from PureFlo PAPRs.

  • Purewipes moist towelettes that clean dirt and perspiration from PureFlo PAPRs.
  • Individually Packaged and Sealed
  • 100 wipes per pack

Specifications

Product Type Disinfectant Wipe
Autoclavable Not Autoclavable
Color White
Content And Storage Individually Packaged and Sealed
Disposable Single-use
Description Purewipes Moist Towelettes that Clean Dirt and Perspiration From PureFlo PAPRs.
For Use With (Application) Clean Dirt and Perspiration from PureFlo PAPRs
For Use With (Equipment) PureFlo PAPR
Packaging Quantity 100/Pk.
Saturation Solution 70% Isopropyl Alcohol
Sterility Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.