Learn More
New England Biolabs, Inc. PURExpress (aa, tRNA) Kit – 10 reactions
SDP
Catalog No. 504351
Encompass
PURExpress (aa, tRNA) Kit is a variation of the PURExpress In vitro Protein Synthesis Kit where the amino acids and tRNA are omitted from the translation mix.
Catalog No. 50-435-1 Supplier New England Biolabs, Inc. Supplier No. E6840S
PURExpress (aa, tRNA) Kit is a variation of the PURExpress In vitro Protein Synthesis Kit where the amino acids and tRNA are omitted from the translation mix.