Learn More
New England Biolabs, Inc. PURExpress delta Ribosome Kit – 10 reactions
SDP
Catalog No. 504304
Encompass
PURExpress delta ribosome kit is a variation of the PURExpress In vitro Protein Synthesis Kit where ribosomes are omitted from the translation mix.
Catalog No. 50-430-4 Supplier New England Biolabs, Inc. Supplier No. E3313S
PURExpress delta ribosome kit is a variation of the PURExpress In vitro Protein Synthesis Kit where ribosomes are omitted from the translation mix.