Learn More
New England Biolabs, Inc. PURExpress Disulfide Bond Enhancer – 50 reactions
SDP
Catalog No. 504350
Encompass
The PURExpress Disulfide Bond Enhancer is a proprietary blend of proteins and buffer components formulated to correctly fold target proteins with multiple disulfide bonds.
Catalog No. 50-435-0 Supplier New England Biolabs, Inc. Supplier No. E6820S
The PURExpress Disulfide Bond Enhancer is a proprietary blend of proteins and buffer components formulated to correctly fold target proteins with multiple disulfide bonds.