Learn More
New England Biolabs, Inc. PURExpress RF123 Kit – 10 reactions
SDP
Supplier:  New England Biolabs, Inc. E6850S
Encompass
PURExpress RF123 kit is a variation of the PURExpress In vitro Protein Synthesis Kit where the release factors 1, 2 and 3 are omitted from the translation mix.
Catalog No. 50-435-2