Learn More
Medchemexpress LLC Purfalcamine 10mg | 1038620-68-6 | 528.62 | C29H33FN8O | 10 MG
SDP
Catalog No. 5000308328
Encompass_Preferred
Purfalcamine is a selective Plasmodium falciparum calcium-dependent protein kinase 1 (PfCDPK1) inhibitor used for antimalarial research. It shows potent enzymatic inhibition (IC50 = 17 nM) and cellular activity (EC50 = 230 nM), causing parasite developmental arrest at the schizont stage. The compound is supplied for research use with characterized purity and composition.
  • Selective PfCDPK1 inhibitor
  • Potent enzymatic inhibition (IC50 17 nM)
  • Demonstrates antimalarial cellular activity (EC50 230 nM)
  • Causes parasite developmental arrest at the schizont stage
  • High reported chemical purity (99.44%)
  • Molecular formula C29H33FN8O and molecular weight 528.62
Catalog No. 50-003-08328 Supplier Medchemexpress LLC Supplier No. HY11701510MG
Purfalcamine is a selective Plasmodium falciparum calcium-dependent protein kinase 1 (PfCDPK1) inhibitor used for antimalarial research. It shows potent enzymatic inhibition (IC50 = 17 nM) and cellular activity (EC50 = 230 nM), causing parasite developmental arrest at the schizont stage. The compound is supplied for research use with characterized purity and composition.
  • Selective PfCDPK1 inhibitor
  • Potent enzymatic inhibition (IC50 17 nM)
  • Demonstrates antimalarial cellular activity (EC50 230 nM)
  • Causes parasite developmental arrest at the schizont stage
  • High reported chemical purity (99.44%)
  • Molecular formula C29H33FN8O and molecular weight 528.62