Learn More
Medchemexpress LLC Purfalcamine 5mg | 1038620-68-6 | 528.62 g/mol | C29H33FN8O | 5 MG
SDP
Catalog No. 5000308401
Encompass_Preferred
Purfalcamine is a research-grade small molecule inhibitor of Plasmodium falciparum calcium-dependent protein kinase 1 (PfCDPK1) with reported antimalarial activity. It is intended for laboratory research use only and is used in biochemical and cellular assays to investigate parasite kinase function.
  • Selective inhibitor of PfCDPK1 for antimalarial research.
  • Reported purity of 99.44% suitable for in vitro assays.
  • Available as a solid (multiple sizes, including 5 mg) and as a 10 mM solution in DMSO.
  • Molecular weight 528.62 g/mol; molecular formula C29H33FN8O.
  • Provided for research use only; not for human or clinical applications.
Catalog No. 50-003-08401 Supplier Medchemexpress LLC Supplier No. HY1170155MG
Purfalcamine is a research-grade small molecule inhibitor of Plasmodium falciparum calcium-dependent protein kinase 1 (PfCDPK1) with reported antimalarial activity. It is intended for laboratory research use only and is used in biochemical and cellular assays to investigate parasite kinase function.
  • Selective inhibitor of PfCDPK1 for antimalarial research.
  • Reported purity of 99.44% suitable for in vitro assays.
  • Available as a solid (multiple sizes, including 5 mg) and as a 10 mM solution in DMSO.
  • Molecular weight 528.62 g/mol; molecular formula C29H33FN8O.
  • Provided for research use only; not for human or clinical applications.