Learn More
Aviva Systems Biology Corp PURG Antibody - N-terminal region (ARP50381_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50381_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PURG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PURG. Alternate names: MGC119274, PURG-A, PURG-BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-60333