Learn More
Perkin Elmer US LLC Disposable Desiccant Kit - Qty. 2
Catalog No. 509052156
Encompass_Preferred
FT-IR spectrometers require dessicant kits to keep their optical paths moisture-free. PerkinElmer recommends that your instrument's desiccant be replaced every six months. Your spectrometer's manual has instructions on how to install the dessicant. Depending on the spectrometer, you will need two to three dessicant kits every six months.
Catalog No. 50-905-2156 Supplier Perkin Elmer US LLC Supplier No. N0171159
FT-IR spectrometers require dessicant kits to keep their optical paths moisture-free. PerkinElmer recommends that your instrument's desiccant be replaced every six months. Your spectrometer's manual has instructions on how to install the dessicant. Depending on the spectrometer, you will need two to three dessicant kits every six months.