Learn More
Perkin Elmer US LLC Radial Purge Extension Window for Optima 3000/3000 SCR/3x00 RL/4300/5300/7300 V
Catalog No. 509043622
Encompass_Preferred
The Radial View Purge Extension Window Tube is a replaceable quartz tube that slides into the mount between the torch and the optics for the radial view.
Catalog No. 50-904-3622 Supplier Perkin Elmer US LLC Supplier No. N0581497
The Radial View Purge Extension Window Tube is a replaceable quartz tube that slides into the mount between the torch and the optics for the radial view.