Learn More
MilliporeSigma™ Supelco™ Purge/Trap B
Catalog No. p-7202910 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
OI Analytical Eclipse 4660, 4560
Catalog No. For Use With (Equipment)
11-100-4301 OI Analytical Eclipse 4660, 4560
1 options
Catalog No. 11-100-4301 Supplier MilliporeSigma™ Supelco™ Supplier No. 24931
Only null left
Add to Cart
Add to Cart
Specific configurations available for a variety of instruments
  • Tubes used in purge and trap analyses generally are packed with multiple beds of adsorbent materials
  • A single tube can trap a broad range of polar and nonpolar, high and low molecular weight compounds
  • Each bed protects the next, increasingly active bed, by preventing compounds from being held so strongly that they cannot be desorbed quickly without decomposition
  • During the purge phase of sampling, lower molecular weight compounds pass through the initial adsorbent beds but are trapped by succeeding beds
  • During desorption, the carrier gas passes through the trap in the reverse direction of purge flow, so that higher molecular weight compounds never come in contact with the stronger (innermost) adsorbents
TRUSTED_SUSTAINABILITY
Specifications
Type Purge/Trap B
For Use With (Application) Industrial Hygiene
For Use With (Equipment) OI Analytical Eclipse 4660, 4560