Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Purge/Trap J BTEXTRAP™

Catalog No. 111010822 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
OI Analytical Eclipse 4660, 4560
Tekmar 3000, 3100
Tekmar Velocity XPT, LSC-1, LSC-2, LSC-2000, 4000, HT-3
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-101-0822 Tekmar Velocity XPT, LSC-1, LSC-2, LSC-2000, 4000, HT-3
11-100-9992 OI Analytical Eclipse 4660, 4560
11-101-0634 Tekmar 3000, 3100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-0822 Supplier MilliporeSigma™ Supelco™ Supplier No. 21064
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specific configurations available for a variety of instruments

  • Tubes used in purge and trap analyses generally are packed with multiple beds of adsorbent materials
  • A single tube can trap a broad range of polar and nonpolar, high and low molecular weight compounds
  • Each bed protects the next, increasingly active bed, by preventing compounds from being held so strongly that they cannot be desorbed quickly without decomposition
  • During the purge phase of sampling, lower molecular weight compounds pass through the initial adsorbent beds but are trapped by succeeding beds
  • During desorption, the carrier gas passes through the trap in the reverse direction of purge flow, so that higher molecular weight compounds never come in contact with the stronger (innermost) adsorbents
TRUSTED_SUSTAINABILITY

Specifications

Connection Type Model 4000: Straight, 12 in., Swagelok Fitting
Type Purge/Trap J
For Use With (Application) Industrial Hygiene
For Use With (Equipment) Tekmar Velocity XPT, LSC-1, LSC-2, LSC-2000, 4000, HT-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.