Learn More
Perkin Elmer US LLC Radial Purge Extension Window for Avio 200 & Optima 2x00/7000 DV/8000
Catalog No. 509043702
Encompass_Preferred
The Radial View Purge Extension Window is a replaceable quartz window that slides into the purge extension between the torch and the optics.
Catalog No. 50-904-3702 Supplier Perkin Elmer US LLC Supplier No. N0690672
The Radial View Purge Extension Window is a replaceable quartz window that slides into the purge extension between the torch and the optics.