Learn More
Andwin Scientific PURGE VALVE ASSEMBLY
SDP
Catalog No. NC1073121
Encompass
The Agilent Purge Valve Long With PTFE Frit is compatible to use with G1310, G1311, G1312, G1376, G2226 pumps.
Catalog No. NC1073121 Supplier Andwin Scientific Supplier No. G131260071
The Agilent Purge Valve Long With PTFE Frit is compatible to use with G1310, G1311, G1312, G1376, G2226 pumps.