Learn More
Glass Expansion Purged Optical Path (POP) Window
SDP
Catalog No. 502703043
Encompass_Preferred
Purged Optical Path (POP) Ceramic‐tipped Interface Kit compatible with the Thermo Fisher Scientific Duo 6000/7000 Series ICP‐OES.The unique design of Glass Expansion’s new POP ceramic‐tipped assembly matches the sensitivity of the standard quartz Radial window, but eliminates the cloudiness and rapid sample build‐up seen with a quartz window (P/N 31‐808‐2677 or 842312051591). A ceramic tip design was also chosen to provide ahigher temperature resistance compared to the quartz options.Replacement window and O-ring ordered as P/N 70-803-1892.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3043 Supplier Glass Expansion Supplier No. 708031571
Purged Optical Path (POP) Ceramic‐tipped Interface Kit compatible with the Thermo Fisher Scientific Duo 6000/7000 Series ICP‐OES.The unique design of Glass Expansion’s new POP ceramic‐tipped assembly matches the sensitivity of the standard quartz Radial window, but eliminates the cloudiness and rapid sample build‐up seen with a quartz window (P/N 31‐808‐2677 or 842312051591). A ceramic tip design was also chosen to provide ahigher temperature resistance compared to the quartz options.Replacement window and O-ring ordered as P/N 70-803-1892.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001