Learn More
MOLEKULE Purgo RX
SDP
Catalog No. NC2403253
Encompass_Preferred
Pūrgo is an air hygiene device that provides continuous air sanitization and supplemental ventilation  for indoor air, removing harmful airborne viruses and other bioaerosols, such as bacteria, fungi, and  pollen from occupied spaces. Pūrgo™ uses patented SteriDuctTM technology that uses safe, solid-state UV-C emitters at the optimum wavelength to eliminate harmful pathogens.Coverage: 4 to 6 air changes/hour in a 3000 ft3 room.
Catalog No. NC2403253 Supplier MOLEKULE Supplier No. PM100A
Pūrgo is an air hygiene device that provides continuous air sanitization and supplemental ventilation  for indoor air, removing harmful airborne viruses and other bioaerosols, such as bacteria, fungi, and  pollen from occupied spaces. Pūrgo™ uses patented SteriDuctTM technology that uses safe, solid-state UV-C emitters at the optimum wavelength to eliminate harmful pathogens.Coverage: 4 to 6 air changes/hour in a 3000 ft3 room.