Learn More
iQ Biosciences Purified C57BL/6 Mouse CD3 Pan T Cells, 5M cells/vial
Supplier:  iQ Biosciences IQBM2T5
Encompass
High-quality, high-viability (typically > 70%) mouse CD3 Pan T cells purified from spleen of C57BL/6 strain of mice. Highly enriched (> 90%) post-selection purity achieved via magnetic isolation. All orders come with an iQ Certificate of Analysis.
Catalog No. NC1985069