Learn More
Biolegend Purified anti-human C5L2
Supplier:  Biolegend 342402
Encompass_Preferred
Purified anti-human C5L2 [1D9-M12]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, WB; Size: 100 ug
Catalog No. NC0574210