Learn More
Biolegend Purified anti-human CD194 (CCR4)
Supplier:  Biolegend 359402
Encompass_Preferred
Purified anti-human CD194 (CCR4) [L291H4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus; Apps: FC; Size: 100 ug
Catalog No. NC1553684