Learn More
Biolegend Purified anti-human TCR α/β
Supplier:  Biolegend 306702
Encompass_Preferred
Purified anti-human TCR α/β [IP26]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. NC1147533