Learn More
iQ Biosciences Purified Cynomolgus Monkey CD14+ Cells, 1M cells/vial
Supplier:  iQ Biosciences IQBMN1CD141
Encompass_Preferred
High-quality, high-viability (> 90%) CD14+ cells purified from cynomolgus macaque peripheral blood mononuclear cells (PBMC) by magnetic isolation. Highly enriched (typically > 80%) post-selection purity achieved via magnetic isolation. Ideal source for cynomolgus monocytes and neutrophils. Used for a wide variety of safety assessment and functional assays. All orders come with an iQ Certificate of Analysis.The following donor requests can be accommodated if inventory is available: - Single or multiple donors for a particular product (SKU). Indicate number of vials from each individual donor - Donor-matched PBMCs
Catalog No. NC1879522