Learn More
Creative Diagnostics Purified exosomes, Lyophilized, 2 x 100 μg, 50-120 nm, Human Plasma, Reconstitute with 100 μL deionized water
Catalog No. NC3566888
Encompass
Purified exosomes, Lyophilized, 2 x 100 μg, 50-120 nm, Human Plasma, Reconstitute with 100 μL deionized water
Catalog No. NC3566888 Supplier Creative Diagnostics Supplier No. DAGA987
Purified exosomes, Lyophilized, 2 x 100 μg, 50-120 nm, Human Plasma, Reconstitute with 100 μL deionized water