Learn More
Hach Company DQ3 Purification Pack
Supplier:  Hach Company 2512201
Encompass
Purification cartridge for Millipore Direct-Q 3 Water System. Pretreatment of tap water by activated carbon, purification by reverse osmosis followed by ion-exchange resin. Package quanitity is one purification pack. One purification pack is required per change.
Catalog No. 50-199-6138