Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Purification PDIV

Catalog No. MP11PN0000 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 x 15 mL Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP11PN0000 10 x 15 mL Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP11PN0000 Supplier MP Biomedicals™ Supplier No. 11PN0000
Add to Cart
Add to Cart

Purification PDIV Tubes used for applications such as gene expression.

  • Contains10 × 15 mL tubes
  • Made up of glass beads, Matrix D, and Matrix B
  • Store at room temperature
TRUSTED_SUSTAINABILITY

Specifications

Bead Type Glass Beads
Content And Storage Store at room temperature
For Use With (Application) Gene expression
Matrix Matrix D and Matrix B
Quantity 10 x 15 mL Tubes
Substrate Glass beads, Matrix D, and Matrix B

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.