Learn More
Biolegend Purified anti-mouse MERTK (Mer)
Supplier:  Biolegend 151502
Encompass_Preferred
Purified anti-mouse MERTK (Mer) [2B10C42]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1527770