Learn More
Biolegend Purified anti-α-Synuclein, 80-96
Supplier:  Biolegend 848302
Encompass_Preferred
Purified anti-α-Synuclein, 80-96 [A15115A]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P, Direct ELISA, WB; Size: 100 ug
Catalog No. NC1822310