Learn More
Biolegend Purified anti-A2M
Catalog No. 502796463
Encompass_Preferred
Purified anti-A2M [A21084F]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: WB, ICFC, IHC-P; Size: 100 μg
Catalog No. 50-279-6463 Supplier Biolegend Supplier No. 649852
Purified anti-A2M [A21084F]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: WB, ICFC, IHC-P; Size: 100 μg