Learn More
Biolegend Purified anti-AGR2
Supplier:  Biolegend 943101
Encompass_Preferred
Purified anti-AGR2 [W19087A]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: WB, ICC, IP; Size: 25 ug
Catalog No. 50-207-5449