Learn More
Biolegend Purified (azide-free) anti-β-Amyloid, 1-42
Supplier:  Biolegend 805503
Encompass_Preferred
Purified (azide-free) anti-β-Amyloid, 1-42 [12F4]; Isotype: Mouse IgG1; Reactivity: Human, Mouse, Rat; Apps: IHC-P, Direct ELISA, WB, IHC-F; Size: 1 mL
Catalog No. 50-207-3943