Learn More
Biolegend Purified (azide-free) anti-β-Amyloid, 1-43
Supplier:  Biolegend 805601
Encompass_Preferred
Purified (azide-free) anti-β-Amyloid, 1-43 [9C4/Amyloid]; Isotype: Mouse IgG1; Reactivity: Human; Apps: IHC-P, WB; Size: 200 uL
Catalog No. 50-207-3947