Learn More
Biolegend Purified anti-Annexin A3
Catalog No. 502866020
Encompass_Preferred
Purified anti-Annexin A3 [O91A5]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, IHC-P; Size: 25 μg
Catalog No. 50-286-6020 Supplier Biolegend Supplier No. 659951
Purified anti-Annexin A3 [O91A5]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, IHC-P; Size: 25 μg