Learn More
Biolegend Purified anti-Annexin A3
Supplier:  Biolegend 659952
Encompass_Preferred
Purified anti-Annexin A3 [O91A5]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, IHC-P; Size: 100 μg
Catalog No. 50-286-5979