Learn More
Biolegend Purified (azide-free) anti-Apo E
Supplier:  Biolegend 803303
Encompass_Preferred
Purified (azide-free) anti-Apo E [D6E10]; Isotype: Mouse IgG1; Reactivity: Human; Apps: WB, IHC-P; Size: 1 mL
Catalog No. 50-208-9905