Learn More
Biolegend Purified (azide-free) anti-Apo E4
Supplier:  Biolegend 811701
Encompass_Preferred
Purified (azide-free) anti-Apo E4 [5A9]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB; Size: 100 uL
Catalog No. 50-207-4024