Learn More
Biolegend Purified (azide-free) anti-Apo E4
Supplier:  Biolegend 811602
Encompass_Preferred
Purified (azide-free) anti-Apo E4 [5B5/ApoE4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA, IHC; Size: 25 uL
Catalog No. 50-207-4023