Learn More
Biolegend Purified (azide-free) anti-APP C-Terminal Fragment
Supplier:  Biolegend 802803
Encompass_Preferred
Purified (azide-free) anti-APP C-Terminal Fragment [C1/6.1]; Isotype: Mouse IgG1; Reactivity: Human, Mouse, Rat; Apps: WB, ICC, IP; Size: 25 uL
Catalog No. NC1509078