Learn More
Biolegend Purified anti-BRST-3
Supplier:  Biolegend 915206
Encompass_Preferred
Purified anti-BRST-3 [B72.3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: IHC-P; Size: 100 ug
Catalog No. 50-207-5204