Learn More
Biolegend Purified anti-CD34
Supplier:  Biolegend 826401
Encompass_Preferred
Purified anti-CD34 [QBEnd/10]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: IHC; Size: 50 uL
Catalog No. 50-207-4228