Learn More
Biolegend Purified anti-CDC73
Supplier:  Biolegend 941702
Encompass_Preferred
Purified anti-CDC73 [W19164B]; Isotype: Rat IgG2b, κ; Reactivity: Human, Mouse; Apps: WB, IP, ICC; Size: 100 ug
Catalog No. 50-207-5438