Learn More
Biolegend Purified (azide-free) anti-E. coli RNA Polymerase β
Supplier:  Biolegend 663905
Encompass_Preferred
Purified (azide-free) anti-E. coli RNA Polymerase β [8RB13]; Isotype: Mouse IgG2b, κ; Reactivity: E. coli, Bordetella, Chlamydiae, Myxococcus, Salmonella, Vibrio; Apps: WB, ELISA, IP; Size: 100 ug
Catalog No. 50-208-7213