Learn More
Biolegend Purified anti-E. coli RNA Sigma S
Supplier:  Biolegend 663705
Encompass_Preferred
Purified anti-E. coli RNA Sigma S [1RS1]; Isotype: Mouse IgG1; Reactivity: E. coli, Klebsiella, Salmonella, Serratia, Bordetella; Apps: WB, ELISA, FA; Size: 25 ug
Catalog No. 50-208-7211