Learn More
Biolegend Purified (azide-free) anti-FMRP
Supplier:  Biolegend 834701
Encompass_Preferred
Purified (azide-free) anti-FMRP [5C2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, IHC-P, ICC; Size: 100 uL
Catalog No. 50-102-9965