Learn More
Biolegend Purified (azide-free) anti-FMRP
Supplier:  Biolegend 834601
Encompass_Preferred
Purified (azide-free) anti-FMRP [6B8/FMRP]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, ICC; Size: 100 uL
Catalog No. 50-102-9964