Learn More
Biolegend Purified anti-Gasdermin-D (N-terminus)
Supplier:  Biolegend 939702
Encompass_Preferred
Purified anti-Gasdermin-D (N-terminus) [W18029B]; Isotype: Rat IgG2b, λ; Reactivity: Human; Apps: WB, IP; Size: 100 ug
Catalog No. 50-207-5409