Learn More
Biolegend Purified anti-GFAP (Cocktail)
Supplier:  Biolegend 835304
Encompass_Preferred
Purified anti-GFAP (Cocktail) [SMI 22]; Isotype: Mouse IgG2b; Reactivity: Human, Sheep, Cow, Dog, Pig, Guinea Pig, Rat, Mouse, Chicken; Apps: IHC; Size: 100 ug
Catalog No. 50-207-4325