Learn More
Biolegend Purified anti-GFAP (Cocktail)
Supplier:  Biolegend 837604
Encompass_Preferred
Purified anti-GFAP (Cocktail) [SMI 26]; Isotype: Mouse IgG1/Mouse IgG2b; Reactivity: Human, Sheep, Cow, Dog, Pig, Rat, Guinea Pig, Rat, Mouse, Chicken; Apps: IHC; Size: 100 ug
Catalog No. 50-207-4414