Learn More
Biolegend Purified anti-Hexokinase II
Supplier:  Biolegend 944001
Encompass_Preferred
Purified anti-Hexokinase II [W19163B]; Isotype: Rat IgG2b, κ; Reactivity: Human, Mouse; Apps: WB; Size: 25 ug
Catalog No. 50-207-5455