Learn More
Biolegend Purified anti-HIF1A
Catalog No. 502721803
Encompass_Preferred
Purified anti-HIF1A [W22058A]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: IHC-P, WB; Size: 25 μg
Catalog No. 50-272-1803 Supplier Biolegend Supplier No. 633351
Purified anti-HIF1A [W22058A]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: IHC-P, WB; Size: 25 μg