Learn More
Biolegend Purified anti-His Tag
Supplier:  Biolegend 362602
Encompass_Preferred
Purified anti-His Tag [J095G46]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat, All species; Apps: WB, FC, ICC; Size: 100 ug
Catalog No. NC1459032